Learn More
Gerstel 10ML CLEAR SCREW CAP VIALS/MPS
SDP
Catalog No. NC9080423
Encompass
10 mL clear screw cap vials for MPS (rounded bottom for transport), Package of 100 Requires magnetic screw caps with septa 093640-092-00 (PolyMag) or 093640-040-00 (metal)
Catalog No. NC9080423 Supplier Gerstel Supplier No. 09364003800
10 mL clear screw cap vials for MPS (rounded bottom for transport), Package of 100 Requires magnetic screw caps with septa 093640-092-00 (PolyMag) or 093640-040-00 (metal)