Learn More
Montana Molecular Borealis Arrestin Assay - B2AR D2040G, CMV, 10mL
SDP
Supplier:  Montana Molecular D2040G010
Encompass_Preferred
Use the B2AR Borealis arrestin assay to measure arrestin signaling kinetics with an unmodified B2AR receptor. Add the receptor and fluorescent sensor to cells and measure robust signals the next day on an automated plate reader or fluorescence imaging system. Compare with cAMP signaling responses using Montana Molecular’s cADDis cAMP assay. These assays are genetically encoded and packaged in BacMam for efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells. Each kit includes the sensor, receptor, GRK, and positive control agonist.
Catalog No. NC2312589