Learn More
Andwin Scientific 10ML DISP PIPETS PS ST 200CS
SDP
Catalog No. NC0567769
Encompass_Preferred
The 10mL Disposable Aspirating Pipets are single-use pipets that are sterile, non-plugged, and nongraduated. These pipets are made from polystyrene, which is a safer material than glass. This product comes in a case of 200.
Catalog No. NC0567769 Supplier Andwin Scientific Supplier No. 414004267
The 10mL Disposable Aspirating Pipets are single-use pipets that are sterile, non-plugged, and nongraduated. These pipets are made from polystyrene, which is a safer material than glass. This product comes in a case of 200.