Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Midwest Scientific 10mL Economy Reagent Reservoirs, 100/pk
SDP

Catalog No. NC1648351
Encompass

Reservoir, 10ml sterile, Individually wrapped, 100/pk

Catalog No. NC1648351 Supplier Midwest Scientific Supplier No. PRSR8P80101S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Reservoir, 10ml sterile, Individually wrapped, 100/pk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.