Learn More
Vmrd Inc Feline Herpesvirus Type 1 (FHV-1/FVR) FITC Conjugate | 10 mL
SDP
Supplier:  Vmrd Inc CJFFVR10ML
Encompass
Anti-Feline Herpesvirus Type 1/Feline Viral Rhinotracheitis (FHV-1/FVR) polyclonal antiserum conjugated to fluorescein isothiocyanate. Feline origin. Useful as an aid in the detection of Feline Viral Rhinotracheitis virus in tissue sections, smears and cultures. Does not stain calicivirus or panleukopenia virus.
Catalog No. NC1666298