Learn More
Chemglass Life Sciences 10mL Flowmeter, Bubble
SDP
Catalog No. CG160701
Encompass_Preferred
10mL Flowmeter, Bubble. Used for the control of carrier gases. Projection at top breaks bubbles. Supplied complete with rubber bulb. Hose connection has a O.D. of 9mm at the middle ring.
Catalog No. CG160701 Supplier Chemglass Life Sciences Supplier No. CG160701
10mL Flowmeter, Bubble. Used for the control of carrier gases. Projection at top breaks bubbles. Supplied complete with rubber bulb. Hose connection has a O.D. of 9mm at the middle ring.