Learn More
Chemglass Life Sciences 10mL Addition Funnel, 14/20 Joint Size, 2mm Glass Stpk, 210mm OAH
SDP
Catalog No. CG170010
Encompass_Preferred
10mL Addition Funnel, 14/20 Joint Size, 2mm Glass Stopcock, 210mm OAH. Cylindrical style addition funnel with a standard taper top outer joint and lower inner joint, both being the same size. With pressure equalizing arm and glass stopcock. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.
Catalog No. CG170010 Supplier Chemglass Life Sciences Supplier No. CG170010
Add to Cart
Add to Cart
10mL Addition Funnel, 14/20 Joint Size, 2mm Glass Stopcock, 210mm OAH. Cylindrical style addition funnel with a standard taper top outer joint and lower inner joint, both being the same size. With pressure equalizing arm and glass stopcock. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.