Learn More
Advalue Technologies Quartz Crucible Cylindrical
SDP
Catalog No. NC0881016
Encompass
FQ-2010 Quartz Crucible - Cylindrical - Capacity 10ml - Diameter 22mm - Height 33mm
Catalog No. NC0881016 Supplier Advalue Technologies Supplier No. FQ2010
FQ-2010 Quartz Crucible - Cylindrical - Capacity 10ml - Diameter 22mm - Height 33mm