Learn More
Chemglass Life Sciences 10mL Addition Funnel, Graduated, 14/20 Joint Size, 2mm Glass Stpk, 210mm OAH
SDP
Catalog No. CG171010
Encompass_Preferred
10mL Addition Funnel, Graduated, 14/20 Joint Size, 2mm Glass Stopcock, 210mm OAH. Cylindrical style addition funnel graduated with a double scale of numbers, one up from the stopcock and one down from the capacity line. With a standard taper top outer joint and lower inner joint, both being the same size, and pressure equalizing arm. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.
Catalog No. CG171010 Supplier Chemglass Life Sciences Supplier No. CG171010
10mL Addition Funnel, Graduated, 14/20 Joint Size, 2mm Glass Stopcock, 210mm OAH. Cylindrical style addition funnel graduated with a double scale of numbers, one up from the stopcock and one down from the capacity line. With a standard taper top outer joint and lower inner joint, both being the same size, and pressure equalizing arm. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.