Learn More
Ace Glass, Inc. 10mL hemispherical fabric mantle, Glas-Col, 32mm max flask diameter, 0.81in inside depth, 1.75in OD, 1.06in height, 30vac max, 20 watts, not rated
SDP
Catalog No. 50977263
Encompass
10mL hemispherical fabric Glas-col mantle, adaptable to unusual shapes and sizes, and will withstand 450 Deg.C. Mantle has a separable 2-wire 3 cord with locking connector. cULus rated. CSA certified.
Catalog No. 50-977-263 Supplier Ace Glass, Inc. Supplier No. 1203502
10mL hemispherical fabric Glas-col mantle, adaptable to unusual shapes and sizes, and will withstand 450 Deg.C. Mantle has a separable 2-wire 3 cord with locking connector. cULus rated. CSA certified.