Learn More
Research Products International Corp 10ml High Capacity Centrifuge Tubes , 10 bgs of 50, 500/pk
SDP
Catalog No. 502252994
Encompass
New 10mL screw cap centrifuge tube are shorter than standard 15ml tubes and allow better sample recovery of contents. Compatible with most 15mL centrifuge rotors, racks, block heaters, etc. and with stand up to 12,000xg maximum RCF. Polypropylene tubes feature printed graduations from 1.5-10ml (0.5ml increments) and flat screw cap.
Printed graduatioms, conical format
Withstand -86°C to 121°C
Sterile, RNase, DNase and Pyrogen free
Screw cap
This item is not returnable. View return policy
Catalog No. 50-225-2994 Supplier Research Products International Corp Supplier No. 248550
New 10mL screw cap centrifuge tube are shorter than standard 15ml tubes and allow better sample recovery of contents. Compatible with most 15mL centrifuge rotors, racks, block heaters, etc. and with stand up to 12,000xg maximum RCF. Polypropylene tubes feature printed graduations from 1.5-10ml (0.5ml increments) and flat screw cap.
Printed graduatioms, conical format
Withstand -86°C to 121°C
Sterile, RNase, DNase and Pyrogen free
Screw cap