Learn More
Phenomenex Inc 10mL Inside Tube
Catalog No. 502399206 Shop All Phenomenex Products
Encompass_Preferred
10mL Inside Tube 6g MgSO4 (Anhydrous),1.5g AnhydrousNaAc, with 10mL
Catalog No. 50-239-9206 Supplier Phenomenex Inc Supplier No. MSMG5052S
10mL Inside Tube 6g MgSO4 (Anhydrous),1.5g AnhydrousNaAc, with 10mL