Learn More
Ace Glass, Inc. 10mL 24/40 Barrett moisture test receiver, jacketed sidearm, glass plug
SDP
Catalog No. 50975342
Encompass_Preferred
10mL Barrett type moisture test receiver with 24/40 standard taper joints for use in determining the water content in petroleum and bituminous material. Features a jacketed side arm and is supplied with 2mm bore glass plug.
Catalog No. 50-975-342 Supplier Ace Glass, Inc. Supplier No. 7745102
10mL Barrett type moisture test receiver with 24/40 standard taper joints for use in determining the water content in petroleum and bituminous material. Features a jacketed side arm and is supplied with 2mm bore glass plug.