Learn More
Trajan Scientific VICI Valco Sample Loop, 10-mL, Cheminert, Stainless Steel (VAL.CSL10K)
Catalog No. 50635006
Encompass
VICI Valco Sample Loop, 10-mL, Cheminert, Stainless Steel, SS Loop With 1/16" Tube Ends - Includes Two SS Nuts and Ferrules (NEW LPP PN: VAL.CSL10K). Thermo Part Number: CH-952987. Made in the USA.
Catalog No. 50-635-006 Supplier Trajan Scientific Supplier No. PAL.CSL10K
VICI Valco Sample Loop, 10-mL, Cheminert, Stainless Steel, SS Loop With 1/16" Tube Ends - Includes Two SS Nuts and Ferrules (NEW LPP PN: VAL.CSL10K). Thermo Part Number: CH-952987. Made in the USA.