Learn More
CELLTREAT Scientific Products 10mL Low Retention Pipette Tips, Racked, Sterile
SDP
Catalog No. NC2204352
Encompass_Preferred
Sterile, accurate liquid transfer of larger volumes of media and solutions. Universal fit is compatible with a range of popular pipettes. More precise than serological pipets for volumes of 1 to 10mL. 5mL tips have molded graduations, with 0.5mL increments.
This item is not returnable. View return policy
Catalog No. NC2204352 Supplier CELLTREAT Scientific Products Supplier No. 229040
Sterile, accurate liquid transfer of larger volumes of media and solutions. Universal fit is compatible with a range of popular pipettes. More precise than serological pipets for volumes of 1 to 10mL. 5mL tips have molded graduations, with 0.5mL increments.