Learn More
S2 Media 10mL MacConkey (MAC) Broth, 16x115mm Polypropylene Flat Bottom Tube, 50/box
Catalog No. 502995896
Encompass_Preferred
MacConkey Broth is a selective and differential liquid medium intended for selection and isolation of enteric gram-negative bacilli, such as coliforms. The original formulation of MacConkey Broth, developed by MacConkey in 1900, contained 0.5% sodium taurocholate (inhibitor) and litmus (indicator). Several iterations were subsequently proposed and in 1905 litmus was replaced with neutral red. In 1953, Childs and Allen replaced neutral red with bromocresol purple after studies demonstrated it was less inhibitory. Sodium taurocholate has also been replaced with oxgall in the current formulation for the inhibition of gram-positive organisms. • Product meets CLSI, ISO, Harmonized USP/EP/JP, and WHO reference methods and performance criteria.• For in vitro diagnostic (IVD) use.• MAC Broth should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5896 Supplier S2 Media Supplier No. 5210B
MacConkey Broth is a selective and differential liquid medium intended for selection and isolation of enteric gram-negative bacilli, such as coliforms. The original formulation of MacConkey Broth, developed by MacConkey in 1900, contained 0.5% sodium taurocholate (inhibitor) and litmus (indicator). Several iterations were subsequently proposed and in 1905 litmus was replaced with neutral red. In 1953, Childs and Allen replaced neutral red with bromocresol purple after studies demonstrated it was less inhibitory. Sodium taurocholate has also been replaced with oxgall in the current formulation for the inhibition of gram-positive organisms. • Product meets CLSI, ISO, Harmonized USP/EP/JP, and WHO reference methods and performance criteria.• For in vitro diagnostic (IVD) use.• MAC Broth should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.