Learn More
Chemglass Life Sciences 10mL Micro Volumetric Flask, Class A, Tolerance: +-0.02mL
SDP
Catalog No. CG160405
Encompass_Preferred
10mL Micro Volumetric Flask, Class A, Tolerance: ¿0.02mL. Class ¿A¿ micro volumetric flask calibrated to contain. Flask has a GPI 13-425 top thread and is supplied complete with an open top screw cap and PTFE faced silicone septa. Flasks have hex base for stability and ¿V¿ shaped interior bottom for total sample retrieval. See MW-102 for replacement caps and MW-104 for replacement septa.
Catalog No. CG160405 Supplier Chemglass Life Sciences Supplier No. CG160405
10mL Micro Volumetric Flask, Class A, Tolerance: ¿0.02mL. Class ¿A¿ micro volumetric flask calibrated to contain. Flask has a GPI 13-425 top thread and is supplied complete with an open top screw cap and PTFE faced silicone septa. Flasks have hex base for stability and ¿V¿ shaped interior bottom for total sample retrieval. See MW-102 for replacement caps and MW-104 for replacement septa.