Learn More
Ace Glass, Inc. 10mL 24/40 Barrett moisture test receiver, glass plug
SDP
Catalog No. 50975341
Encompass_Preferred
10mL Barrett type moisture test receiver with 24/40 standard taper joints for use in determining the water content in petroleum and bituminous material. Supplied with 2mm bore glass plug.
Catalog No. 50-975-341 Supplier Ace Glass, Inc. Supplier No. 774502
10mL Barrett type moisture test receiver with 24/40 standard taper joints for use in determining the water content in petroleum and bituminous material. Supplied with 2mm bore glass plug.