Learn More
Chemglass Life Sciences Distilling Receiver, 10mL, Moisture Test, Dean-Stark, 14/20 Joint Size, Approx. 250mm Height x 80mm Width
SDP
Supplier:  Chemglass Life Sciences CG125901
Encompass_Preferred
Distilling Receiver, 10mL, Moisture Test, Dean-Stark, 14/20 Joint Size, Approx. 250mm Height x 80mm Width. Similar to CG-1258 but with the addition of an overflow tube between the uptake tube and the graduated receiver.
Catalog No. CG125901