Learn More
Analytical Sales & Services 10mL PEEK Prep Sample Loop
Catalog No. 5000804774
Encompass
10mL PEEK Prep Sample Loop, 1/8" OD x 2mm ID, 5/16-24 Thread Port Configuration\n\nQty/Pkg: EACH
Catalog No. 50-008-04774 Supplier Analytical Sales & Services Supplier No. 430550234
10mL PEEK Prep Sample Loop, 1/8" OD x 2mm ID, 5/16-24 Thread Port Configuration\n\nQty/Pkg: EACH