Learn More
Chemglass Life Sciences 10mL Peptide Synthesis Vessel, Solid Phase, Medium Frit, GL 14 Thread
SDP
Catalog No. CG186101
Encompass_Preferred
Solid phase peptide synthesis vessel having a medium or coarse porosity fritted glass resin support. Top of the vessel has a GL thread and is supplied complete with a PTFE lined PBT screw cap. Side port and lower PTFE stopcock have a 2mm bore with the arms being 8mm I.D. Vessels are available in sizes other than those listed. Please call for details.
Catalog No. CG186101 Supplier Chemglass Life Sciences Supplier No. CG186101
Solid phase peptide synthesis vessel having a medium or coarse porosity fritted glass resin support. Top of the vessel has a GL thread and is supplied complete with a PTFE lined PBT screw cap. Side port and lower PTFE stopcock have a 2mm bore with the arms being 8mm I.D. Vessels are available in sizes other than those listed. Please call for details.