Learn More
Bellco Glass, Inc. 10mL Pipet Individually
SDP
Catalog No. 502330256
Encompass_Preferred
All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets.Specifications Peeltoopen plastic wrappers are the ideal option for eliminating particulate proven benefit over traditional paper wrapper Wrappers reduce static cling versus other brands of plastic wraps Can be wiped with alcohol without smearing ink or penetrating paper 100 Individually Wrapped Case Quantity 800
Catalog No. 50-233-0256 Supplier Bellco Glass, Inc. Supplier No. 229210A00
All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets.Specifications Peeltoopen plastic wrappers are the ideal option for eliminating particulate proven benefit over traditional paper wrapper Wrappers reduce static cling versus other brands of plastic wraps Can be wiped with alcohol without smearing ink or penetrating paper 100 Individually Wrapped Case Quantity 800