Learn More
Bellco Glass, Inc. 10mL Pipette Tips, Ovation, Graduated, VistaBulk, Non-sterile
SDP
Catalog No. 502330387
Encompass_Preferred
A superior fit for your MLA & Ovation Pipettes. Use genuine VistaLab pipette tips to maximize the exceptional performance and benefits of Ovation and MLA pipettes. All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time.Macro Volume•Offer improved accuracy over serological pipets•Tip length allows better access to the bottom of test tubes, reagent bottles, flasks and other vessels•5mL macro tips are graduated at 0.1mL increments•Available sterile and non-sterile in a variety of packaging options: Box, VistaClear Box, VistaBulk , and VistaTip •Tips are manufactured from 100% virgin polypropylene and can be autoclaved•VistaClear Box Sterile, filtered are certified RNase/DNase free & Non-Pyrogenic
Catalog No. 50-233-0387 Supplier Bellco Glass, Inc. Supplier No. 451900202
A superior fit for your MLA & Ovation Pipettes. Use genuine VistaLab pipette tips to maximize the exceptional performance and benefits of Ovation and MLA pipettes. All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time.Macro Volume•Offer improved accuracy over serological pipets•Tip length allows better access to the bottom of test tubes, reagent bottles, flasks and other vessels•5mL macro tips are graduated at 0.1mL increments•Available sterile and non-sterile in a variety of packaging options: Box, VistaClear Box, VistaBulk , and VistaTip •Tips are manufactured from 100% virgin polypropylene and can be autoclaved•VistaClear Box Sterile, filtered are certified RNase/DNase free & Non-Pyrogenic