Learn More
Triforest Enterprises 10mL Autoclaveable Polycarbonate Serum Vial
Supplier:  Triforest Enterprises RPC0010
Encompass_Preferred
TriForest PC Serum Vials are ideal for sampling, storage, production and packaging of steam sterilized biologicals and shipment of reagents and buffer solutions. All vials have lot number traceability, provided non-sterile.
Catalog No. NC2401709