Learn More
Integra Biosciences Corp 10 mL Polyproyplene Reservoir, 30 individually wrapped per case
SDP
Catalog No. NC1768198
Encompass
Users benefit by reusing the sturdy base and tossing the disposable insert. Reservoirs nest inside each other, making it possible to get twice as many reservoirs in half the space of other products on the market, reducing inventory space requirements and shipping costs. Unlike traditional reservoirs that have hard to see graduations molded directly into the plastic, INTEGRA reservoirs fit into a reusable base with bold, crisp, clearly visible graduation markings. The unique design leads to more accurate reagent measurements, no over pouring, and less waste.
Catalog No. NC1768198 Supplier Integra Biosciences Corp Supplier No. 4336
Users benefit by reusing the sturdy base and tossing the disposable insert. Reservoirs nest inside each other, making it possible to get twice as many reservoirs in half the space of other products on the market, reducing inventory space requirements and shipping costs. Unlike traditional reservoirs that have hard to see graduations molded directly into the plastic, INTEGRA reservoirs fit into a reusable base with bold, crisp, clearly visible graduation markings. The unique design leads to more accurate reagent measurements, no over pouring, and less waste.