Learn More
S2 Media 10mL Potato Dextrose Broth (PDB) in 15mL Conical Tube, 50/box
Catalog No. 502995957
Encompass_Preferred
10mL Potato Dextrose Broth (PDA) is an enrichment broth used for the cultivation of yeasts and molds. The low pH of this medium can inhibit some bacterial growth. • Product meets AOAC, APHA, ASM, CLSI, Harmonized USP/EP/JP, and SM-Dairy reference methods and performance criteria.• PDB should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5957 Supplier S2 Media Supplier No. 5300A
10mL Potato Dextrose Broth (PDA) is an enrichment broth used for the cultivation of yeasts and molds. The low pH of this medium can inhibit some bacterial growth. • Product meets AOAC, APHA, ASM, CLSI, Harmonized USP/EP/JP, and SM-Dairy reference methods and performance criteria.• PDB should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.