Learn More
Chemglass Life Sciences 10mL Powder Addition Funnel, 14/20 Joint Size, Auger A
SDP
Catalog No. 50873276
Encompass_Preferred
10mL Powder Addition Funnel, 14/20 Joint Size, Auger A. Facilitates the delivery of free-flowing, granular solids into reactions. Pressure equalizing arm is equipped with a vented stopper for air-free operations and also permits access into the delivery stem for easy clean-out. One piece, easily removed grooved PTFE auger delivers powders at a uniform rate.F313
Catalog No. 50-873-276 Supplier Chemglass Life Sciences Supplier No. CG172301
10mL Powder Addition Funnel, 14/20 Joint Size, Auger A. Facilitates the delivery of free-flowing, granular solids into reactions. Pressure equalizing arm is equipped with a vented stopper for air-free operations and also permits access into the delivery stem for easy clean-out. One piece, easily removed grooved PTFE auger delivers powders at a uniform rate.F313