Learn More
Chemglass Life Sciences 10mL Single Neck Round Bottom Flask (RBF) , 14/20 Outer Joint, Side Tubulation
SDP
Catalog No. CG1514A02
Encompass_Preferred
10mL Single Neck Round Bottomed Flask, 14/20 Outer Joint, Side Tubulation. Round bottom, HEAVY WALL flask with standard taper outer joint. Side tubulation accepts a CG-1040 rubber fitment for insertion of thermometers or bleed tubes. Supplied complete with one CG-1040 rubber fitting.
Catalog No. CG1514A02 Supplier Chemglass Life Sciences Supplier No. CG1514A02
10mL Single Neck Round Bottomed Flask, 14/20 Outer Joint, Side Tubulation. Round bottom, HEAVY WALL flask with standard taper outer joint. Side tubulation accepts a CG-1040 rubber fitment for insertion of thermometers or bleed tubes. Supplied complete with one CG-1040 rubber fitting.