Learn More
Chemglass Life Sciences 10mL Recovery Flask Insert, Anodized ORANGE Aluminum, OptiTherm (for use with OPT100)
SDP
Catalog No. 501531377
Encompass_Preferred
10mL Recovery Flask Insert, Anodized ORANGE Aluminum, OptiTherm (for use with OPT100). Optitherm blocks provide a SAFE and cost effective solution for replacing your existing oil baths and heating mantles. The innovative PTFE shield on the unit reduces the exterior exposed surface temperature by as much as 100C. For example, the PTFE shield would be approximately 80C on a 100mL flask that is heated to 180C. The OPT100 is used for 100mL round bottom flasks.
Catalog No. 50-153-1377 Supplier Chemglass Life Sciences Supplier No. CG1992R010
10mL Recovery Flask Insert, Anodized ORANGE Aluminum, OptiTherm (for use with OPT100). Optitherm blocks provide a SAFE and cost effective solution for replacing your existing oil baths and heating mantles. The innovative PTFE shield on the unit reduces the exterior exposed surface temperature by as much as 100C. For example, the PTFE shield would be approximately 80C on a 100mL flask that is heated to 180C. The OPT100 is used for 100mL round bottom flasks.