Learn More
Sarstedt Inc Test Tube Round Bottom Plain 15.3 X 92 mm 10 mL Screw Cap Polypropylene Tube (1,000/pk)
Catalog No. NC9585761 Shop All Sarstedt Inc. Products
Encompass_Preferred
10ml Screw Cap Tube 15.3x92mm Round Bottom Polypropylene Cap Assembled Sterile
Catalog No. NC9585761 Supplier Sarstedt Inc Supplier No. 60.610.001
10ml Screw Cap Tube 15.3x92mm Round Bottom Polypropylene Cap Assembled Sterile