Learn More
Sarstedt Inc 10ML SC TUBE 15.3X92RB G/CS500
Supplier:  Sarstedt Inc 62.610.201
Encompass_Preferred
10ml Screen Printed Screw Cap Tube 15.3x92mm Round Bottom Polypropylene Graduations and Writing Block Cap Assembled Sterile
Catalog No. NC9746490