Learn More
Sarstedt Inc 10mL Screen Printed Screw Cap Tube, 1000/Case
Supplier:  Sarstedt Inc 62.9924.293
Encompass_Preferred
10mL Screen Printed Screw Cap Tube 16x100mm Conical Bottom Polypropylene Graduations and Writing Block Neutral Cap Enclosed
Catalog No. NC9697894