Learn More
Sarstedt Inc 10ml Screw Cap Tube, Conical Bottom, 500/Pack
Supplier:  Sarstedt Inc 60.9921.821
Encompass_Preferred
10ml Screw Cap Tube 16x97mm Conical Bottom Skirted Polystyrene Neutral Cap
Catalog No. NC9022594