Learn More
Alkali Scientific 10mL Screw Top Headspace Vial
SDP
Catalog No. 503027444
Encompass_Preferred
The ASI10mL Screw Top Headspace Vial is designed for precision efficiency and convenience in chromatography applications These pre-assembled vials include 18mm magnetic precision open-top screw caps with blue PTFE/white silicone septa ensuring a secure fit while reducing preparation time Proper cap and septa selection are essential for maintaining sample integrity and preventing contamination making these vials a dependable choice for laboratories requiring accuracy and consistency Applications Alkali Scientifics screw top headspace vials are ideal for gas chromatography (GC) and high-performance liquid chromatography (HPLC) particularly in headspace sampling techniques used to analyze volatile compounds Common applications include Environmental Analysis Detecting contaminants in water air and soil samples Food and Beverage Testing Identifying aroma compounds in food spices and beverages Pharmaceutical Quality Control Ensuring the purity and consistency of pharmaceutical formulat
Catalog No. 50-302-7444 Supplier Alkali Scientific Supplier No. GCSC010
The ASI10mL Screw Top Headspace Vial is designed for precision efficiency and convenience in chromatography applications These pre-assembled vials include 18mm magnetic precision open-top screw caps with blue PTFE/white silicone septa ensuring a secure fit while reducing preparation time Proper cap and septa selection are essential for maintaining sample integrity and preventing contamination making these vials a dependable choice for laboratories requiring accuracy and consistency Applications Alkali Scientifics screw top headspace vials are ideal for gas chromatography (GC) and high-performance liquid chromatography (HPLC) particularly in headspace sampling techniques used to analyze volatile compounds Common applications include Environmental Analysis Detecting contaminants in water air and soil samples Food and Beverage Testing Identifying aroma compounds in food spices and beverages Pharmaceutical Quality Control Ensuring the purity and consistency of pharmaceutical formulat