Learn More
USA Scientific Inc 10 mL serological pipet, indiv. wrapped, sterile
Catalog No. NC1283704
Encompass
10 mL serological pipet, 0.1 mL graduations, standard tip, individually wrapped, sterile, 200/case
Catalog No. NC1283704 Supplier USA Scientific Inc Supplier No. 56607180
10 mL serological pipet, 0.1 mL graduations, standard tip, individually wrapped, sterile, 200/case