Learn More
Bellco Glass, Inc. 10mL Serological Pipet Individually Wrapped
SDP
Catalog No. 502329770
Encompass_Preferred
All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934Bi-directional (positive and negative) graduations are printed in bold easy-to-read inkPremium material barrier plugColor coded stripes and packaging enable quick volume identificationPrinted indicator arrow for quickly locating negative graduations when using a pipet controller Pack Description50 Individually Wrapped/BagCase Quantity200SterileYesNon-pyrogenicYesPrimary ColorClearSecondary ColorOrangePrimary MaterialPolystyreneMaximum Volume10mLGraduatedYesGraduation Increments (Subdivisions)1/10mLRNase/DNase FreeYes
Catalog No. 50-232-9770 Supplier Bellco Glass, Inc. Supplier No. 229010B00
All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934Bi-directional (positive and negative) graduations are printed in bold easy-to-read inkPremium material barrier plugColor coded stripes and packaging enable quick volume identificationPrinted indicator arrow for quickly locating negative graduations when using a pipet controller Pack Description50 Individually Wrapped/BagCase Quantity200SterileYesNon-pyrogenicYesPrimary ColorClearSecondary ColorOrangePrimary MaterialPolystyreneMaximum Volume10mLGraduatedYesGraduation Increments (Subdivisions)1/10mLRNase/DNase FreeYes