Learn More
USA Scientific Inc 10mL serological , 0.1 mL grad
Catalog No. 502745297
Encompass
10 mL serological pipet, 0.1 mL graduations, standard tip, str, 25/sleeve, 500/case
Catalog No. 50-274-5297 Supplier USA Scientific Inc Supplier No. 56607107
10 mL serological pipet, 0.1 mL graduations, standard tip, str, 25/sleeve, 500/case