Learn More
USA Scientific Inc 10 mL serological pipet, sterile
Catalog No. NC9868325
Encompass
10 mL serological pipet, standard tip orifice, 0.1 mL graduations, individually wrapped, sterile, 50/sleeve, 200/case
Catalog No. NC9868325 Supplier USA Scientific Inc Supplier No. 10710810
10 mL serological pipet, standard tip orifice, 0.1 mL graduations, individually wrapped, sterile, 50/sleeve, 200/case