Learn More
Bellco Glass, Inc. 10ml Serological Pipette, Individually Wrapped, sterile 200/pk, 800/cs
SDP
Catalog No. 502330252
Encompass_Preferred
10ml Serological Pipette with 3.0ml negative graduation, 1/10 ml graduations, Individually Plastic-plastic Wrapped, sterile, 50/bag, 200/pk, 800/cs. Plastic/plastic packaging permits peel-off function that provides user-friendly easy opening and aseptic pipette removal, reduced contamination risk and particulate-free opening. Fine line graduation printing contributes to accuracy of at least +/- 2%. Reduced contamination risk and particulate-free openingFine line graduation printing contributes to accuracy of at least +/- 2%Negative graduations provide increased volume versatilityReverse (ascending and descending) graduations on 5, 10, 25 and 50 mL pipettesSynthetic filter and color-coded according to international conventionLot number and best before date on each individual wrap
Catalog No. 50-233-0252 Supplier Bellco Glass, Inc. Supplier No. 451400246
10ml Serological Pipette with 3.0ml negative graduation, 1/10 ml graduations, Individually Plastic-plastic Wrapped, sterile, 50/bag, 200/pk, 800/cs. Plastic/plastic packaging permits peel-off function that provides user-friendly easy opening and aseptic pipette removal, reduced contamination risk and particulate-free opening. Fine line graduation printing contributes to accuracy of at least +/- 2%. Reduced contamination risk and particulate-free openingFine line graduation printing contributes to accuracy of at least +/- 2%Negative graduations provide increased volume versatilityReverse (ascending and descending) graduations on 5, 10, 25 and 50 mL pipettesSynthetic filter and color-coded according to international conventionLot number and best before date on each individual wrap