Learn More
Bellco Glass, Inc. 10ml Serological Pipette, Sterile, 20 per bulk bag, 500/pk, 1500/cs
SDP
Catalog No. 502329518
Encompass_Preferred
10ml Serological Pipette with 3.0ml negative graduation, 1/10 ml graduations, Sterile, 20 per bulk bag, 500/pk, 1500/cs. Use for liquid transfer. bulk packaging serologcials offer the same high quality serological pipettes, but in a more environment friendly way.
Catalog No. 50-232-9518 Supplier Bellco Glass, Inc. Supplier No. 451400252
10ml Serological Pipette with 3.0ml negative graduation, 1/10 ml graduations, Sterile, 20 per bulk bag, 500/pk, 1500/cs. Use for liquid transfer. bulk packaging serologcials offer the same high quality serological pipettes, but in a more environment friendly way.