Learn More
CEM Corporation 10 mL Pressure Vessels, Pyrex
SDP
Catalog No. 501877896
Encompass
10 mL pressure rated vessels, pack of 100. The 10 mL pressure vessels have a working volume of 0.2-7.0 mL for synthesis applications, in the Discover SP, and 1.0-4.0 mL for digestion applications, in the Discover SP-D Clinical, and are rated up to 600 psi.
Catalog No. 50-187-7896 Supplier CEM Corporation Supplier No. 908035
10 mL pressure rated vessels, pack of 100. The 10 mL pressure vessels have a working volume of 0.2-7.0 mL for synthesis applications, in the Discover SP, and 1.0-4.0 mL for digestion applications, in the Discover SP-D Clinical, and are rated up to 600 psi.