Learn More
Ace Glass, Inc. 10ML SPEC GRAV BTL COMP 10ML SPEC GRAV BTL COMP
SDP
Catalog No. 50971878
Encompass_Preferred
Complete 10mL Specific Gravity bottle set. Approximate assembled height is 168mm. Includes conical shape flask, 25mm non-mercury immersion thermometer with 10/18 joint and 14 to 37 degree C range, and cap with 5/12 standard taper joint.
Catalog No. 50-971-878 Supplier Ace Glass, Inc. Supplier No. 541508
Complete 10mL Specific Gravity bottle set. Approximate assembled height is 168mm. Includes conical shape flask, 25mm non-mercury immersion thermometer with 10/18 joint and 14 to 37 degree C range, and cap with 5/12 standard taper joint.