Learn More
Integra Biosciences Corp 10 ml Poly SureFlo™ Poly Reservoirs, pk/200
SDP
Catalog No. NC1840770
Encompass
Users benefit by reusing the sturdy base and tossing the disposable insert. Reservoirs nest inside each other, making it possible to get twice as many reservoirs in half the space of other products on the market, reducing inventory space requirements
This item is not returnable. View return policy
Catalog No. NC1840770 Supplier Integra Biosciences Corp Supplier No. 4372
Users benefit by reusing the sturdy base and tossing the disposable insert. Reservoirs nest inside each other, making it possible to get twice as many reservoirs in half the space of other products on the market, reducing inventory space requirements