Learn More
S2 Media 10mL Tetrathionate (TT) Broth Base, 16x115mm Polypropylene Flat Bottom Tube, 50/box
Catalog No. 502995956
Encompass_Preferred
Tetrathionate (TT) Broth Base is a selective enrichment medium used with an iodine-iodide solution for the cultivation of Salmonella spp. from clinical specimens, foods, and other materials of sanitary importance. Mueller demonstrated the effectiveness of Tetrathionate Broth for enriching typhoid and paratyphoid bacilli while inhibiting coliform organisms. Kauffmann increased the number of positive isolates by modifying Mueller's broth.• Product meets AOAC, BAM, CLSI, EP, SM-Dairy, SMWW, and USP-NF performance criteria.• Tetrathionate Broth should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5956 Supplier S2 Media Supplier No. 5198
Tetrathionate (TT) Broth Base is a selective enrichment medium used with an iodine-iodide solution for the cultivation of Salmonella spp. from clinical specimens, foods, and other materials of sanitary importance. Mueller demonstrated the effectiveness of Tetrathionate Broth for enriching typhoid and paratyphoid bacilli while inhibiting coliform organisms. Kauffmann increased the number of positive isolates by modifying Mueller's broth.• Product meets AOAC, BAM, CLSI, EP, SM-Dairy, SMWW, and USP-NF performance criteria.• Tetrathionate Broth should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.