Learn More
Chemglass Life Sciences 10 mL Transport Tube, Short, Sterile, Caps On, USP VI Polypropylene, HDPE Screw Cap, 50/Pack
SDP
Catalog No. 502733402
Encompass_Preferred
Sterile 10 mL short transport tubes for safe storage, transport, and freezing of biological and clinical samples. Tubes are molded from USP VI-grade polypropylene and fitted with HDPE screw caps that provide leak-resistant sealing suitable for air transport; they are free of RNase, DNase, pyrogens, and endotoxins.
  • Sterile by electron beam sterilization with SAL = 10^-6.
  • Constructed from USP VI polypropylene with HDPE caps.
  • Leak-resistant screw caps withstand negative pressure up to 95 kPa for air transport safety.
  • Packaged 50 per pack and 1,000 per case.
  • Self-standing design for stable placement on lab benches.
  • Suitable for storage, transport, and freezing of samples in medical, hospital, environmental, food, and research laboratories.
  • DNase-free, RNase-free, pyrogen-free, and endotoxin-free.
  • Temperature range approximately -196°C to 121°C.
Catalog No. 50-273-3402 Supplier Chemglass Life Sciences Supplier No. CGN474210S
Add to Cart
Add to Cart
Sterile 10 mL short transport tubes for safe storage, transport, and freezing of biological and clinical samples. Tubes are molded from USP VI-grade polypropylene and fitted with HDPE screw caps that provide leak-resistant sealing suitable for air transport; they are free of RNase, DNase, pyrogens, and endotoxins.
  • Sterile by electron beam sterilization with SAL = 10^-6.
  • Constructed from USP VI polypropylene with HDPE caps.
  • Leak-resistant screw caps withstand negative pressure up to 95 kPa for air transport safety.
  • Packaged 50 per pack and 1,000 per case.
  • Self-standing design for stable placement on lab benches.
  • Suitable for storage, transport, and freezing of samples in medical, hospital, environmental, food, and research laboratories.
  • DNase-free, RNase-free, pyrogen-free, and endotoxin-free.
  • Temperature range approximately -196°C to 121°C.