Learn More
Chemglass Life Sciences 10mL Transport Tubes, with Caps on, Sterile, 50/pk, 1000/cs
SDP
Catalog No. 502733403
Encompass_Preferred
Transport tubes are used in multiple sectors of the laboratory market, including medical, hospital, environmental, independent testing, food testing, and biological research labs. Used to store, transport, or freeze samples.USP VI Polypropylene Tubes, HDPE CapsSelf-standing: the vial placed on the lab workbench steadily even without a baseElectron Beam Sterilization, SAL=10-6: safe, fast, environment-friendly, no chemical residues100,000-level Clean Workshop Production, DNAse-free, RNAse-free, pyrogen-free, endotoxin-freeTemperature Range: Approx.-196°C to 121°CExcellent sealing performance, no leakage of negative pressure 95Kpa, ensuring safe and effective air transportationRNase and pyrogen freeElectron beam sterilization, SAL=10-6
Catalog No. 50-273-3403 Supplier Chemglass Life Sciences Supplier No. CGN474210
Add to Cart
Add to Cart
Transport tubes are used in multiple sectors of the laboratory market, including medical, hospital, environmental, independent testing, food testing, and biological research labs. Used to store, transport, or freeze samples.USP VI Polypropylene Tubes, HDPE CapsSelf-standing: the vial placed on the lab workbench steadily even without a baseElectron Beam Sterilization, SAL=10-6: safe, fast, environment-friendly, no chemical residues100,000-level Clean Workshop Production, DNAse-free, RNAse-free, pyrogen-free, endotoxin-freeTemperature Range: Approx.-196°C to 121°CExcellent sealing performance, no leakage of negative pressure 95Kpa, ensuring safe and effective air transportationRNase and pyrogen freeElectron beam sterilization, SAL=10-6