Learn More
Chemglass Life Sciences Tube, Vacuum Hydrolysis, 6mL Capacity, 10mm Tube OD x 150mm Tube Length
SDP
Supplier:  Chemglass Life Sciences CG402503
Encompass_Preferred
Tube, Vacuum Hydrolysis, 6mL Capacity, 10mm Tube OD x 150mm Tube Length. Tube is useful in protein hydrolysis, amino acid separations, freeze drying or any application under vacuum.
Catalog No. CG402503