Learn More
Andwin Scientific 10ML VACUTAINER EDTA LAV PK100
SDP
Catalog No. 501125965
Encompass_Preferred
Vacutainer Venous Blood Collection Tubes Plastic EDTA tube 16x100mm x 10.0mL.BD Vacutainer Lavender BD Hemogard closure. See-through label. 
This item is not returnable. View return policy
Catalog No. 50-112-5965 Supplier Andwin Scientific Supplier No. BD366643
Vacutainer Venous Blood Collection Tubes Plastic EDTA tube 16x100mm x 10.0mL.BD Vacutainer Lavender BD Hemogard closure. See-through label. 