Learn More
Gerstel 10ML VIALS CLEAR 100/PK
SDP
Catalog No. NC0058082
Encompass
10 mL clear vials for MPS (rounded bottom for transport), Package of 100 Requires magnetic crimp caps with septa (093640-061-00)
Catalog No. NC0058082 Supplier Gerstel Supplier No. 09364000500
10 mL clear vials for MPS (rounded bottom for transport), Package of 100 Requires magnetic crimp caps with septa (093640-061-00)