Learn More
Analytical Sales & Services 10mm, 12 x 32, 2mL Amber Glass Round Bottom Wide Mouth Screw Vials
Catalog No. 5000806688
Encompass
Advantage™ 10mm, 12 x 32, 2mL Volume Amber Glass Round Bottom Wide Mouth Screw Vials - for Shimadzu and Waters Autosamplers
Catalog No. 50-008-06688 Supplier Analytical Sales & Services Supplier No. 12320ACASE
Advantage™ 10mm, 12 x 32, 2mL Volume Amber Glass Round Bottom Wide Mouth Screw Vials - for Shimadzu and Waters Autosamplers